Infusion filters set, Intrapur® Inline
Leverandør: B. Braun
4099842N
BRAU4099842NEA 9100 NOK
BRAU4099842N
Infusion filters set, Intrapur® Inline
Infusjoner
Ready-to-use set with infusion filter.
  • PVC free
  • Retention of bacteria and particles
  • High throughput
Specifically designed for use during chemotherapy.