Infusion filter, Intrapur® Plus
Leverandør: B. Braun
4183916
BRAU4183916EA 20850 NOK
BRAU4183916
Infusion filter, Intrapur® Plus
Infusjoner
Intrapur plus offers reliable air venting regardless of position with connection lines at both sides of the filter with Luer lock fittings.
  • High throughput
  • PVC free
  • Positively charged membrane
NB: Should not be used for more than 96 hours.