Sterican® Single-Use Needles
Leverandør: B. Braun
4657624B
BRAU4657624BEA 89 NOK
BRAU4657624B
Sterican® Single-Use Needles
Sprøytenåler
Stainless chromium-nickel steel needle with extremely smooth surface and light silicone coating, transparent Luer lock PP hub.
  • Colour coded hub, conforms to ISO 6009
  • Latex- and PVC-free
  • Tri-bevelled for minimal pain upon puncture (long bevel)
Sertifiseringer: ISO 7864, DIN 13097
Forpakning: Individually, sterile packed. 100 needles (20 blocks of 5) per box.