Gauze, Askina®
Leverandør: B. Braun
9030409W
BRAU9030409WEA 2325 NOK
BRAU9030409W
Gauze, Askina®
Førstehjelpsutstyr Bandasjer
Absorbent gauze for making compresses and swabs, as an ointment or medication carrier.
  • 80 cm wide, folded into 8 layers to a width of 10 cm
  • Bleached 100% without chlorine
  • Highly absorbent
  • Can be cut to size
Sertifiseringer: DIN 61630 - VM 20