Bandages, Askina® Elast
Leverandør: B. Braun
9048120
BRAU9048120EA 275 NOK
BRAU9048120
Bandages, Askina® Elast
Førstehjelpsutstyr Bandasjer
Elastic, wide-mesh fixation bandage made from 70% viscose and 30% polyamide. For all types of fixation dressings and light support bandages.
  • Very comfortable thanks to high natural fibre content
  • Good adhesion of the individual turns of the bandage thanks to creped woven structure
  • Approx. 130% lengthways elasticity
  • Can be sterilised in an autoclave