Fixation plasters, Askina® silk
Leverandør: B. Braun
9080120N
BRAU9080120NEA 1700 NOK
BRAU9080120N
Fixation plasters, Askina® silk
Førstehjelpsutstyr Bandasjer
Silk plasters for the fixation of all types of wound dressing, e.g. Askina® Pad, Askina® gauze compresses. Also suitable for the attachment of catheters, tubes and puncture sets.
  • Reliable adhesion on different surfaces
  • Skin-friendly acrylate adhesive
  • Very easy to tear, removable with little pain