Insulin syringes, Omnifix® 40 Solo
Leverandør: B. Braun
9161309V
BRAU9161309VEA 425 NOK
BRAU9161309V
Insulin syringes, Omnifix® 40 Solo
Sprøyter Sprøyter, medisinsk
Omnifix® syringes are single-use insulin syringes for U-40.
  • Latex-free plug
  • Safe piston stop avoids loss of medicine
  • Highly transparent barrel