Fine dosage syringes, Omnifix®-F Duo
Leverandør: B. Braun
9161465V
BRAU9161465VEA 640 NOK
BRAU9161465V
Fine dosage syringes, Omnifix®-F Duo
Sprøyter Sprøyter, medisinsk
These fine dosage syringes are made of PP/PS. Plunger stopper with double sealing ring for slow aspiration and injection made of natural latex free synthetic material.
  • Highly transparent barrel with permanent marking
  • Safe plunger backstop
  • Easy aspiration up to maximum volume
  • Minimal residual volume
  • Latex free/PVC free
Sertifiseringer: According to EN ISO 7886-1, EN ISO 7864 and DIN 13097