Printer cleaning kit
Leverandør: Brady
018556
BRDY018556EA 285 NOK
BRDY018556
Printer cleaning kit
Papirservietter Rengjøringssett og svamper
This cleaning kit for printers includes: Instruction sheet, five cleaning cards soaked in IPA and the material safety data sheet.