Labels for BBP3x printers, type B-549
Leverandør: Brady
117865 117860 117861 117862 117863 117864
BRDY117865EA 3750 NOK
BRDY117865 BRDY117860 BRDY117861 BRDY117862 BRDY117863 BRDY117864
Labels for BBP3x printers, type B-549
Etiketter
Cold temperature tape permanently adheres to surfaces at temperatures as low as −10 °F.
  • Ideal for use in any cold storage warehouse, cooler or freezer
  • Not recommended for plotter cutting