i7100 Vial Label Applicator
Leverandør: Brady
151156
BRDY151156EA 61275 NOK
BRDY151156
i7100 Vial Label Applicator
Printere Label Printers
Automate vial tube identification with the Vial Label Applicator accessory that prints and applies labels on vial tubes in a few seconds.
Works only with the 300 and 600 dpi Peel models of the BradyPrinter i7100 Industrial Label Printers.
Leveringsinformasjon: The accessories for the wraptor printer applicator includes cleaning kits, memory cards, additional hardware, shippping materials, and stylus.