Solvent and Chemical Resistant Polyester Labels with Ribbon for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Leverandør: Brady
170774 170887
BRDY170774EA 855 NOK
BRDY170774 BRDY170887
Solvent and Chemical Resistant Polyester Labels with Ribbon for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Etiketter
High-performance polyester (B-488) withstands numerous solvents and variable temperatures when applied to various surfaces, including stainless steel and polypropylene.
  • Specialized – Use B-488 high-performance polyester label material for lab applications with numerous solvents and variable temps
  • Durable – Labels resist chemicals, abrasions, corrosion and humidity and have a high/low temperature performance of –40 to +160 °C (–40 to +320 °F)
  • Adhesive – Made with a permanent acrylic adhesive for tight bonding onto flat, smooth surfaces
Brady AuthenticMore Product Information