Label Printer EMEA, M511
Leverandør: Brady
170970
BRDY170970EA 6450 NOK
BRDY170970
Label Printer EMEA, M511
Printere Label Printers
Work smarter. Label larger with the M511. Print bigger, clearer messages. Go edge to edge. Design and print on the fly, or a PC. Plus, share files, import data and connect up to 5 users simultaneously.
  • Monocolour
  • Ideal for larger applications - print up to 38,10 mm labels
  • Less waste - go edge to edge with text, graphics and barcodes
  • Longer battery life with an internal Li-ION battery - 8+ hours of work; 1000 labels on a single charge
  • More durable - resists 1,8 m drops, shocks, 115 kg crushes and blowing sand and dust (MIL-STD-810G)
  • Prints anywhere -
More Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Grensesnitt Mobile device connected, Peripheral
Label Continuous and die-cut
Modell M511
Trykkmetode Thermal transfer
Trykkhastighet 1000 max. labels/day
Oppløsning 300 dpi