Temperature Indicating Labels, Go or No Go
Leverandør: Brady
195843
BRDY195843EA 370 NOK
BRDY195843
Temperature Indicating Labels, Go or No Go
Temperatur indikatorstrips
Reversible temperature indicating labels are part of the VisAlert® Series from Brady®, which is designed to provide immediate visual indication of water or heat exposure.
  • Large temperature range for applications requiring temperature monitoring between 0 and 100 °C
  • Strong permanent acrylic adhesive for any surface
  • Made of sub-surface printed Polyester
Cost-effective solution for temperature checks during maintenance or for trouble spotting during research and development. Extremely low profile is ideal for components, industrial and householdMore Product Information