Reversible temperature indicator labels
Leverandør: Brady
195844
BRDY195844EA 370 NOK
BRDY195844
Reversible temperature indicator labels
Temperatur indikatorstrips
These reversible temperature indicator labels designed to display temperatures of food and medical supply packaging or of components during research, repair and maintenance.
  • Changes colour at designated temperatures
  • Temperature ranges between 0 to 100 °C
  • Highly customisable
  • Permanent acrylic adhesive
Colour: Red and black.