Spill berm, QuickBerm® rigid lock, EG-5
Leverandør: Brady
198532
BRDY198532EA 21100 NOK
BRDY198532
Spill berm, QuickBerm® rigid lock, EG-5
Materialer for håndtering av søl
The extreme durable Rigid-Lock QuickBerm® provided immediate, secure deployment and spill-readiness. The built-in Rigid-Lock wall supports maximise usable space inside the berm and greatly reduce any tripping hazard on the berm perimeter.
  • Made of PVC coated fabric
  • Black in colour
The high performance material withstands a broad range of chemicals including petroleum, oils, grease, and most acids and a broad UV resistance - durable enough for extended wear.