Over laminating tape
Leverandør: Brady
266081
BRDY266081EA 5325 NOK
BRDY266081
Over laminating tape
Tape
Protect printed messages with this clear over laminating tape.
  • Two-year average outdoor durability in temperatures from −40 to +150 °C
  • Minimum application temperature: 10 °C
Length: 50,0 m