SPC® Absorbents SitePoint Chemical Maintenance Kit
Leverandør: Brady
309080
BRDY309080EA 15000 NOK
BRDY309080
SPC® Absorbents SitePoint Chemical Maintenance Kit
Materialer for håndtering av søl
Spill kit.
  • Loaded SitePoint mobile unit with suitable products for cleaning up general spills
  • Features two, 200 mm wheels for mobility and anchor points for fixed installation
  • Fix items to backboard using self-tapping screws or bolts
  • Suitable for inside or outdoor use
Ideal to move spill control equipment near specific risks. Features two rubber wheels for easy trolley movement to wherever it is required. Perfect during maintenance activities, at construction sites and other temporary locations. Includes three anchor points on the base, allowing theMore Product Information