BradyJet J4000-EU-BWSPWID Label Printer, Colour, with Product and Wire ID Software
Leverandør: Brady
310389
BRDY310389EA 61550 NOK
BRDY310389
BradyJet J4000-EU-BWSPWID Label Printer, Colour, with Product and Wire ID Software
Printere Label Printers
This industry inkjet workhorse creates up to 5000 impactful full-colour labels per day up to 203,20 mm wide on continuous or die-cut material.
  • Full colour
  • Long-lasting
  • 5000 labels per day
Ideal for a wide variety of applications in safety, including lean/5S, lockout tagout, instructional or safety chemical labelling for handling, storage or transport, as well as lab settings. Contains the colour options you need to create ANSI, GHS, ISO, BS5609 and arc flash compliant labels. Streamline storage by only needing to stock white label material and printMore Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Datamaskinkrav PC
Tilkoblinger USB; ethernet
Støttet material Continuous and die-cut
Modell J4000-EU-BWSPWID
Strømforsyning AC adaptor
Skriver Full Colour
Trykkmetode Inkjet
Trykkoppløsning 4800 dpi
Trykkhastighet 5000 max. labels/day
Trykkbredde 209,55 mm (max.)