Rapido™ Wire Marking Tags for M710, M610 and M611, Bulk
Leverandør: Brady
312140 312139 312137 312141 312138
BRDY312140EA 4475 NOK
BRDY312140 BRDY312139 BRDY312137 BRDY312141 BRDY312138
Rapido™ Wire Marking Tags for M710, M610 and M611, Bulk
Etiketter
Rapido™ Wiremarking Sleeves are fast and easy to apply. Simply squeeze the edges of the material together, thread the wire through the holes, and pull away the wire to release the marker.
  • Provides high tear-, smudge- and abrasion-resistance
  • Matte finish
Applications include data and telecommunications labelling, and wire and cable labelling.
Forpakning: 2500 tags per box.