Label Printer, AZERTY, M610
Leverandør: Brady
317792
BRDY317792EA 11800 NOK
BRDY317792
Label Printer, AZERTY, M610
Printere Label Printers
Get the durability your work demands.
  • Design and print labels using a keypad or desktop software
  • Work longer with an enhanced Gen3 Li-ION battery – print 4500 labels on a full charge
  • Easily import and export label files, lists and graphics to the printer through USB 2.0 port (Type A and B) or USB flash drive
  • Print labels at 50,80 mm per second print speed
  • Get added durability with a printer that is resistant to military-grade shocks (MIL-STD-810G) and 1,80 m drops
  • Operate your printer in −18 to +46 °C temperatures
Do more with enhanced capabilitiesMore Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Label Continuous and die-cut
Modul Stand-alone and peripheral
Skriver Monocolour
Trykkmetode Thermal transfer
Trykkoppløsning 300 dpi
Trykkhastighet 750 (recommended usage per day)