Labelling pen, quick drying
Leverandør: Brady
335091
BRDY335091EA 455 NOK
BRDY335091
Labelling pen, quick drying
Penner Permanent Markers
These labelling pens use a quick drying ink.
The permanent ink is specially formulated to resist oil, solvents, abrasion and fading.