Label printer, BSP61
Leverandør: Brady
710949 711040
BRDY710949EA 72950 NOK
BRDY710949 BRDY711040
Label printer, BSP61
Printere Label Printers
The BSP™61 Print and apply system offers a fully automatic labelling solution anywhere along the production line. It combines a thermal transfer label printer with an automatic applicator and is designed to consistently print and accurately position and apply labels in almost all applications.
  • Print speed is 250 mm/s
  • Internal memory capacity is 64 mb with RAM/flash of 8 mb
  • Monocolour
  • Designed to handle up to 7000 labels per day
It remove the problems of automating label application to ensure reliable tracking and tracing with no cutting into the liner,More Product Information