Absorbent pillows, ALLWICK Universal
Leverandør: Brady
813792
BRDY813792EA 3200 NOK
BRDY813792
Absorbent pillows, ALLWICK Universal
Materialer for håndtering av søl
These absorbent pillows absorb spills and leaking fluids in tight spaces and under persistent drip sources.
  • Absorbs a variety of liquids including water, coolants, solvents, oils, antifreeze, etc
  • Ideal under machines or in tight spaces