Mobile container spill kit
Leverandør: Brady
813882 813883
BRDY813882EA 8250 NOK
BRDY813882 BRDY813883
Mobile container spill kit
Materialer for håndtering av søl
These wheeled spill kits are comes in a polyethylene container with hinged lid. Designed for medium sized spills of industrial liquids including oils, water, solvents, coolants and chemical hazardous liquids.
  • Yellow in colour
Container is weather-resistant and protects the sorbents against dirt, moisture and damage. Hinged lid provides easy access to the products. Wheels allow excellent mobility indoors and outdoors.