Spill kit, spill truck
Leverandør: Brady
813888 813889
BRDY813888EA 43675 NOK
BRDY813888 BRDY813889
Spill kit, spill truck
Materialer for håndtering av søl
Spill Truck spill kits are ideal for larger spills.
  • Lid provides easy access to the products
  • Wheels allow excellent mobility indoors and outdoors
  • Weather and chemical resistant container protects sorbents from moisture, dirt and damage
Leveringsinformasjon: Kit contains: 150 pads (400×500 mm), 36 socs (Ø 76×1220 mm), 20 socs (Ø 76×3660 mm), 16 pillows (430×480 mm), 50 wipes (300×380 mm), 1 pair of gloves, 1 pair of goggles and 30 disposal bags.