Compact circuit breaker lockout
Leverandør: Brady
836597
BRDY836597EA 270 NOK
BRDY836597
Compact circuit breaker lockout
Sikkerhetslåser
Compact universal design to lock out a large range of single, double and triple-pole circuit breakers.
Place lockout over circuit breaker handle, tighten set screw and attach padlock and tag.