Repositionable Multi-Purpose Vinyl Cloth Labels B-498 CleanLift® for M7 Printers
Leverandør: Brady
M7-17-498
BRDYM7-17-498EA 1350 NOK
BRDYM7-17-498
Repositionable Multi-Purpose Vinyl Cloth Labels B-498 CleanLift® for M7 Printers
Etiketter
Permanent or repositionable, once applied the B-498 CleanLift® Series Vinyl Cloth remains in place, until it must be changed.
  • Adhesive and cloth backing offers excellent holding power, yet allows for clean removal and repositioning
  • Sticks to any clean, dry surface
  • Specially formulated topcoat for excellent print quality
  • Semi-gloss finish for crisp, clean look
Labels are ideal for asset tracking, barcode labelling, circuit board labelling, component and equipment labelling, data and telecommunications labelling, electrical labelling, generalMore Product Information