Pipe marker, H₂S
Leverandør: Brady
N004308
BRDYN004308EA 220 NOK
BRDYN004308
Pipe marker, H₂S
Skilt
Individual pipe markers on map with perforated arrows, with pictogram.
  • Rubber-based permanent adhesive
  • Polyester with polyester overlaminate material
  • Colour: Black on yellow