Pipe Marker, Steam
Leverandør: Brady
N005604
BRDYN005604EA 220 NOK
BRDYN005604
Pipe Marker, Steam
Skilt
Individual pipe markers on map with perforated arrows, without pictogram.
  • Polyester with polyester overlaminate
  • Permanent rubber-based adhesive
  • Coloir: Black on grey
Forpakning: Total quantity per pack - five marker(s)/card.