Dropping bottles, glass, with pipette
Leverandør: Brand
124529 124539 124633 124639
BRND124529EA 305 NOK
BRND124529 BRND124539 BRND124633 BRND124639
Dropping bottles, glass, with pipette
Flasker Dråpeflasker
Soda-lime glass, clear or amber.
  • Good chemical and physical properties
  • For use in applications that do not involve high thermal stresses
  • Replaceable NS glass pipette with rubber suction bulb
Amber to protect light-sensitive substances.