Strømningsindikator, rørehjul
Leverandør: Brand
134080
BRND134080EA 535 NOK
BRND134080
Strømningsindikator, rørehjul
Flytindikatorer
Transparent styrene acrylonitrile (SAN) useful device to optimise cooling water flow during distillation and reduce water consumption.