Trakter med kort hals
Leverandør: Brand
145505 145507 145515 145520 145525 145530 145535 145540 145545 145550 145560
BRND145505EA 100 NOK
BRND145505 BRND145507 BRND145515 BRND145520 BRND145525 BRND145530 BRND145535 BRND145540 BRND145545 BRND145550 BRND145560
Trakter med kort hals
Trakter Filtreringstrakter Filtertrakter
Borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With smooth internal walls. Flare angle 60°. Funnel lip is ground ore fire-polished.