Ice buckets, octagonal, with lid
Leverandør: Brand
156100
BRND156100EA 3750 NOK
BRND156100
Ice buckets, octagonal, with lid
Isbeholdere
Polyurethane foam.
  • Excellent insulating properties
  • Sturdy, octagonal design for improved stability
  • Tapered side with molded handles
  • Suitable for chemical disinfection
  • Operating temperature −196 °C to +95 °C
Ideal for storing samples, these buckets maintain hot or cold temperatures of bottles, test tubes, solutions, and cultures. Suitable for use with dry ice, water ice, salt solution. For bench-top cold storage.