Fargebeholder, Hellendahl, utvidet
Leverandør: Brand
474400
BRND474400EA 2100 NOK
BRND474400
Fargebeholder, Hellendahl, utvidet
Krukker Fargetrau
Made of transparent PMP. Complete with lid.
These staining trough hold up to 16 slides (76×26 mm), 8 pairs back to back.