Krukker, med skrulokk
Leverandør: Brand
62310
BRND62310EA 3225 NOK
BRND62310
Krukker, med skrulokk
Krukker Bokser, generell bruk
PP, translucent.
  • Conical shape
  • Excellent chemical resistance
  • Excellent temperature resistance
Ideal for sampling, transportation and storage.