Centrifuge tubes
Leverandør: Brand
778013
BRND778013EA 2650 NOK
BRND778013
Centrifuge tubes
Rør Sentrifugerør
Duran® borosilicate glass.
  • Withstands forces up to 3500×g
Conical bottom, beaded rim, ungraduated.