Microcentrifuges tubes, with attached cap, BIO-CERT® PCR QUALITY, Brand®
Leverandør: Brand
780420
BRND780420EA 495 NOK
BRND780420
Microcentrifuges tubes, with attached cap, BIO-CERT® PCR QUALITY, Brand®
Rør Mikrosentrifugerør
The disposable microtubes with 1,5 ml nominal volume are made from high-quality PP. The quality level BIO-CERT® PCR QUALITY ensures the required cleanliness for PCR-setups, storage of PCR samples and other applications with high cleanliness requirements. This includes absence of DNA, RNase, DNase, pyrogenes and PCR inhibitors.
  • High cleanliness for demanding applications
  • Tight sealing snap lid
  • Can be opened and closed with one hand
They are available with an easy-to-open snap-on lid in versions with or without lid locking. Lid locking preventsMore Product Information