Prøveøser
Leverandør: Brand
90438 90448 90454 90462
BRND90438EA 3250 NOK
BRND90438 BRND90448 BRND90454 BRND90462
Prøveøser
Prøvetakere Prøveøser
Sampling dipper with reinforced rim and spout.
  • PTFE handle with steel core
Optional extension rod for increased handle length.