Body Showers with Eye/Face Shower and Bowl, Modular
Leverandør: BROEN-LAB
17.556.009
BROE17.556.009EA 37425 NOK
BROE17.556.009
Body Showers with Eye/Face Shower and Bowl, Modular
Førstehjelpsutstyr Dusjer
Body shower with eye/face shower and bowl, modular. In brass and SS with chemical resistant BROEN-LAB Polycoat with self-draining shower head.
  • Professional solutions for employee safety
  • Easy to mount
  • Anti-limescale design and material
  • Improved functionality with long lifetime
  • Ideal for pharma industries
With built-in FLOWFIX for regulation of water flow (16 L/min) and stainless steel bowl with integrated outlet for eye wash.
Min. working pressure: 1,5 bar.