Biotin kits, HOOK™
Leverandør: G Bioscienes
BS-16 BS-10 BS-03 BS-01 BS-02 BS-04 BS-05 BS-06 BS-07 BS-09 BS-11 BS-12 BS-13 BS-14 BS-17 BS-18 BS-19 BS-08
BS-16EA 13325 NOK
BS-16 BS-10 BS-03 BS-01 BS-02 BS-04 BS-05 BS-06 BS-07 BS-09 BS-11 BS-12 BS-13 BS-14 BS-17 BS-18 BS-19 BS-08
Biotin kits, HOOK™
Systemer for proteinkobling og -merking
HOOK™ Biotin Reagent that conjugates to proteins through amines, sulfhydryls, carboxyls or carbohydrates. The amine and sulfhydryl coupling HOOK™ Biotin Reagents couple directly to the protein through their reactive groups, however the carboxyl coupling HOOK™ Biotin Reagents require a carbodiimide crosslinker and the carbohydrate coupling HOOK™ Biotin Reagents require carbohydrate oxidation before coupling.
  • Optimizer Buffer™ for improved coupling efficiency
  • SpinOUT™ gel filtration columns for rapid (<10 minutes) purification
  • Biotin assay reagents to determine level of biotin incorporation
  • Labels 1 to 10 mg protein/reaction
  • Suitable for 10 coupling reactions
The kits include EDC as the carbodiimide crosslinker in the carboxyl coupling kits and sodium meta-periodate for carbohydrate oxidation in the carbohydrate coupling kits. Kit also contains a specific Optimizer Buffer™ that provides the optimal reaction conditions for each HOOK™ Biotin Reagent.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Vert