Anti-ATF6 Rabbit Polyclonal Antibody

Leverandør: Boster Bio
A00655
BSBTA00655EA 6475 NOK
BSBTA00655
Anti-ATF6 Rabbit Polyclonal Antibody
Antistoffer
Rabbit IgG polyclonal antibody for Cyclic AMP-dependent transcription factor ATF-6 alpha(ATF6) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Type: Primary
Antigen: ATF6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR