Anti-IgM Antibody (Biotin)

Leverandør: Boster Bio
BA1022-0.25
BSBTBA1022-0.25EA 0 NOK
BSBTBA1022-0.25
Anti-IgM Antibody (Biotin)
Antistoffer
Type: Secondary
Antigen: IgM
Clonality:
Clone:
Conjugation: Biotin
Epitope:
Host:
Isotype:
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR