Anti-IgM Antibody (FITC (Fluorescein Isothiocyanate))

Leverandør: Boster Bio
BA1116-0.5
BSBTBA1116-0.5EA 0 NOK
BSBTBA1116-0.5
Anti-IgM Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
Type: Secondary
Antigen: IgM
Clonality:
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host:
Isotype:
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR