Mouse CCL19/MIP-3 Beta PicoKine; ELISA Kit

Leverandør: Boster Bio

EK0457
BSBTEK0457EA 7575 NOK
BSBTEK0457
Mouse CCL19/MIP-3 Beta PicoKine; ELISA Kit
Assay ELISAs

Sandwich High Sensitivity ELISA kit for Quantitative Detection of Mouse CCL19/MIP-3 beta.


  • Sensitivity, Detect single digit picogram level of target protein
  • Specificity, Thoroughly tested for cross-reactivity with family members
  • Stability, Guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, On-demand technical support directly from ELISA experts


Detect Mouse Mip-3 Beta/Ccl19 with <10 pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Mip-3 Beta/Ccl19 tissue specificity: Highly expressed by dendritic cells in mesenteric and peripheral lymph nodes. Significant expression in spleen (T cell zone or periarteriolar lymphatic sheath) and Peyer patches. Low expression in thymus.


96wells/kit, with removable strips.

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Primary antibody reactivity Mouse
Analysetype Sandwich
Vert

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR