Human AFP/Alpha 1 Fetoprotein PicoKine; ELISA Kit

Leverandør: Boster Bio

EK0651
BSBTEK0651EA 7575 NOK
BSBTEK0651
Human AFP/Alpha 1 Fetoprotein PicoKine; ELISA Kit
Assay ELISAs

Sandwich High Sensitivity ELISA kit for Quantitative Detection of Human AFP.


  • Sensitivity, Detect single digit picogram level of target protein
  • Specificity, Thoroughly tested for cross-reactivity with family members
  • Stability, Guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, On-demand technical support directly from ELISA experts


Detect Human Alpha-Fetoprotein/AFP with <10 pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Alpha-Fetoprotein/AFP tissue specificity: Plasma. Synthesised by the fetal liver and yolk sac.


96wells/kit, with removable strips.

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR