PicoKine Human CXCL4/PF4; ELISA Kit

Leverandør: Boster Bio

EK0726
BSBTEK0726EA 7575 NOK
BSBTEK0726
PicoKine Human CXCL4/PF4; ELISA Kit
Assay ELISAs

Sandwich high sensitivity ELISA kit for quantitative detection of human CXCL4/PF4.


  • Sensitivity, detect single digit picogram level of target protein
  • Specificity, thoroughly tested for cross-reactivity with family members
  • Stability, guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, on-demand technical support directly from ELISA experts


Detect human Cxcl4/PF4 with <15pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Cxcl4/PF4 tissue specificity:


96wells/kit, with removable strips.

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR