ELISA Kit, Human AGRP/agouti-related PicoKine

Leverandør: Boster Bio

EK1427
BSBTEK1427EA 7575 NOK
BSBTEK1427
ELISA Kit, Human AGRP/agouti-related PicoKine
Assay ELISAs

Sandwich high sensitivity ELISA kit for quantitative detection of human AGRP/agouti-related.


  • Sensitivity, detect single digit picogram level of target protein
  • Specificity, thoroughly tested for cross-reactivity with family members
  • Stability, guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, on-demand technical support directly from ELISA experts


Detect human AGRP with <4 pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, serum, plasma (heparin, EDTA, citrate) and urine. This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. AGRP tissue specificity: expressed primarily in the adrenal gland, subthalamic nucleus, and hypothalamus, with a lower level of expression occurring in testis, lung, and kidney.


96 wells/kit, with removable strips.

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR