Anti-Cathepsin B Polyclonal Antibody

Leverandør: Boster Bio
PB9046
BSBTPB9046EA 6475 NOK
BSBTPB9046
Anti-Cathepsin B Polyclonal Antibody
Antistoffer
Polyclonal antibody for Cathepsin B/CTSB detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. Cathepsin B/CTSB information: Molecular Weight: 37822 MW; Subcellular Localization: Lysosome. Melanosome. Secreted, extracellular space . Identified by mass spectrometry in melanosome fractions from stage I to stage IV.

Type: Primary
Antigen: Cathepsin B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR