Anti-Hsp90 Alpha Polyclonal Antibody

Leverandør: Boster Bio
PB9089
BSBTPB9089EA 6475 NOK
BSBTPB9089
Anti-Hsp90 Alpha Polyclonal Antibody
Antistoffer
Polyclonal antibody for HSP90/HSP90AA1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. HSP90/HSP90AA1 information: Molecular Weight: 84660 MW; Subcellular Localization: Cytoplasm. Melanosome. Cell membrane. Identified by mass spectrometry in melanosome fractions from stage I to stage IV.

Type: Primary
Antigen: Hsp90 Alpha
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR